Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lztfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033322.2 | NP_201579.1 | 3 | 308 | ||
GenBank mRNA | AJ297352.1 | - | 3 | 304 | ||
AK029671.1 | - | 3 | 340 | |||
AK030068.1 | - | 3 | 320 | |||
AK041091.1 | - | 3 | 341 | |||
AK046515.1 | - | 3 | 297 | |||
AK054003.1 | - | 3 | 314 | |||
AK081529.1 | - | 1 | 93 | |||
AK145263.1 | - | 3 | 317 | |||
AK147312.1 | - | 2 | 252 | |||
AK157770.1 | - | 3 | 315 | |||
BC026840.1 | - | 3 | 308 | 4165152 | ||
BC037045.1 | - | 3 | 308 | |||
BC098224.1 | - | 3 | 313 | 1263280 |