Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080819.1 | NP_001074288.1 | 6, 7 | 2260 | ||
GenBank mRNA | AK142143.1 | - | 5, 6 | 1442 | ||
BC082554.1 | - | 1492 | 30622048 |