Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dach2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142570.1 | NP_001136042.1 | 6, 7 | 1386 | ||
NM_001289732.1 | NP_001276661.1 | 9, 10 | 1185 | |||
NM_001289733.1 | NP_001276662.1 | 8, 9 | 1103 | |||
NM_001289734.1 | NP_001276663.1 | 9, 10 | 1254 | |||
NM_033605.2 | NP_291083.1 | 7, 8 | 1425 | |||
GenBank mRNA | AF257217.1 | - | 7, 8 | 1424 | ||
AK033050.1 | - | 9, 10 | 1186 | |||
AK038687.1 | - | 2, 3 | 199 | |||
AK041446.1 | - | 8, 9 | 1092 | |||
AK042401.1 | - | 9, 10 | 1189 | |||
AK045154.1 | - | 7, 8 | 1428 | |||
BC059233.1 | - | 6, 7 | 1375 | 6406445 | ||
GQ247884.1 | - | 4, 5 | 564 |