Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dach2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142570.1 | NP_001136042.1 | 5 | 1181 | ||
NM_001289732.1 | NP_001276661.1 | 8 | 980 | |||
NM_001289733.1 | NP_001276662.1 | 7 | 898 | |||
NM_001289734.1 | NP_001276663.1 | 8 | 1049 | |||
NM_033605.2 | NP_291083.1 | 6 | 1220 | |||
GenBank mRNA | AF257217.1 | - | 6 | 1219 | ||
AK033050.1 | - | 8 | 981 | |||
AK041446.1 | - | 7 | 887 | |||
AK042401.1 | - | 8 | 984 | |||
AK045154.1 | - | 6 | 1223 | |||
BC059233.1 | - | 5 | 1170 | 6406445 | ||
GQ247884.1 | - | 3 | 359 |