Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163660.1 | NP_001157132.1 | 3 | 248 | ||
NM_027280.3 | NP_081556.3 | 5 | 458 | |||
GenBank mRNA | AF343352.1 | - | 5 | 295 | ||
AF358134.1 | - | 5 | 374 | |||
AK076164.1 | - | 5 | 449 | |||
AK082367.1 | - | 5 | 446 | |||
AK166564.1 | - | 5 | 461 | |||
BC034838.1 | - | 5 | 432 | 1429943 | ||
DV662736.1 | - | 3 | 228 |