Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hemgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053149.2 | NP_444379.1 | 3 | 409 | ||
GenBank mRNA | AF269248.1 | - | 3 | 409 | ||
AK076587.1 | - | 4 | 394 | |||
AK077692.1 | - | 3 | 252 | |||
AK156536.1 | - | 3 | 337 | |||
AK171890.1 | - | 3 | 328 | |||
AK171906.1 | - | 3 | 458 | |||
AK172421.1 | - | 3 | 328 | |||
AY699986.1 | - | 3 | 251 | |||
BC050798.1 | - | 4 | 388 | 6743819 |