Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013616.2 | NP_001013637.1 | 8 | 1156 | ||
GenBank mRNA | AK033396.1 | - | 8 | 1136 | ||
AK049238.1 | - | 8 | 1136 | |||
BC026912.1 | - | 1 | 380 | |||
BC138965.1 | - | 8 | 1087 | 8861987 |