Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053169.2 | NP_444399.2 | 4 | 1203 | ||
GenBank mRNA | AF449496.1 | - | 4 | 954 | ||
AK053139.1 | - | 4 | 1017 | |||
AK078902.1 | - | 2 | 539 | |||
BC052821.1 | - | 5 | 905 | 30057830 |