Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053169.2 | NP_444399.2 | 5, 6 | 1344 | ||
GenBank mRNA | AF220134.1 | - | 1, 2 | 62 | ||
AF449496.1 | - | 5, 6 | 1095 | |||
AK053139.1 | - | 5, 6 | 1158 | |||
AK078902.1 | - | 3, 4 | 680 |