Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033373.1 | NP_203537.1 | 3 | 543 | ||
GenBank mRNA | AF102849.1 | - | 3 | 543 | ||
AK036901.1 | - | 3 | 558 | |||
AK036954.1 | - | 3 | 559 | |||
AK145247.1 | - | 3 | 571 | |||
BC003472.1 | - | 3 | 578 | |||
BC019972.1 | - | 3 | 537 | 4187035 |