Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178589.3 | NP_848704.1 | 2, 3 | 1198 | ||
GenBank mRNA | AF322069.1 | - | 2, 3 | 756 | ||
AK033529.1 | - | 2, 3 | 1174 | |||
AK043823.1 | - | 2, 3 | 1200 | |||
AK134704.1 | - | 2, 3 | 1169 | |||
AY043489.1 | - | 2, 3 | 756 | |||
BC016420.1 | - | 2, 3 | 1147 | 4220624 |