Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172098.1 | NP_001165569.1 | 16 | 2296 | ||
NM_052973.2 | NP_443205.1 | 18 | 2548 | |||
GenBank mRNA | AF307777.1 | - | 18 | 2532 | ||
AK140447.1 | - | 16 | 2284 | |||
AK165908.1 | - | 16 | 1980 | |||
BC138055.1 | - | 16 | 2247 | 8861074 | ||
BC145127.1 | - | 16 | 2247 | 9053675 | ||
EF685152.1 | - | 19 | 2449 |