Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033324.2 | NP_201581.2 | 6 | 1818 | ||
GenBank mRNA | AB086857.1 | - | 6 | 1818 | ||
AK048056.1 | - | 5 | 1619 | |||
AK086233.1 | - | 5 | 2010 | |||
BC051013.1 | - | 4 | 2004 | |||
BC062919.1 | - | 6 | 1838 | 6833280 | ||
BC066118.1 | - | 6 | 1865 | 6400561 |