Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srd5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053188.2 | NP_444418.1 | 4, 5 | 795 | ||
GenBank mRNA | AB049456.1 | - | 4, 5 | 795 | ||
AK138946.1 | - | 4, 5 | 736 | |||
BC125510.1 | - | 4, 5 | 779 | 40130025 |