Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC11A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000617.2 | NP_000608.1 | 10 | 991 | ||
NM_001174125.1 | NP_001167596.1 | 10 | 983 | |||
NM_001174126.1 | NP_001167597.1 | 10 | 991 | |||
NM_001174127.1 | NP_001167598.1 | 10 | 986 | |||
NM_001174128.1 | NP_001167599.1 | 10 | 986 | |||
NM_001174129.1 | NP_001167600.1 | 10 | 1251 | |||
NM_001174130.1 | NP_001167601.1 | 9 | 966 | |||
NR_033421.1 | 10 | 911 | ||||
NR_033422.1 | 10 | 986 | ||||
XM_005268911.3 | XP_005268968.1 | 10 | 1043 | |||
XM_005268912.4 | XP_005268969.1 | 9 | 1168 | |||
XM_011538404.2 | XP_011536706.1 | 10 | 1367 | |||
XM_011538405.2 | XP_011536707.1 | 10 | 975 | |||
XM_017019355.1 | XP_016874844.1 | 10 | 1251 | |||
XM_017019356.1 | XP_016874845.1 | 10 | 1181 | |||
GenBank mRNA | AB004857.1 | - | 10 | 934 | ||
AB062284.1 | - | 10 | 908 | |||
AF046997.1 | - | 10 | 944 | |||
AF064484.1 | - | 10 | 934 | |||
AJ493662.1 | - | 10 | 983 | |||
AK094735.1 | - | 10 | 1251 | |||
AK128640.1 | - | 10 | 883 | |||
AK296445.1 | - | 9 | 945 | |||
AK315987.1 | - | 10 | 897 | |||
AK316507.1 | - | 9 | 922 | |||
BC002592.2 | - | 10 | 938 | 3160522 | ||
BC100014.1 | - | 10 | 959 | 5300266 | ||
CK000292.1 | - | 6 | 442 | |||
KT455015.1 | - | 10 | 947 | |||
L37347.1 | - | 7 | 684 |