Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033573.2 | NP_291051.1 | 2 | 725 | ||
GenBank mRNA | AF273750.1 | - | 2 | 746 | ||
AK078394.1 | - | 2 | 745 | |||
BC027831.1 | - | 2 | 758 | |||
BC052072.1 | - | 2 | 725 | 5717482 | ||
BC082784.1 | - | 2 | 743 | 4191155 |