Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168256.1 | NP_001161728.1 | 8 | 638 | ||
NM_001168257.1 | NP_001161729.1 | 7 | 563 | |||
NM_001168258.1 | NP_001161730.1 | 3 | 363 | |||
NM_001168259.1 | NP_001161731.1 | 4 | 130 | |||
NM_144805.2 | NP_659054.2 | 8 | 506 | |||
GenBank mRNA | AK078864.1 | - | 4 | 131 | ||
BC019416.1 | - | 8 | 468 | 5151159 | ||
BX511925.1 | - | 3 | 90 | |||
CT010303.1 | - | 7 | 436 | |||
CV561368.1 | - | 7 | 505 | |||
CX564997.1 | - | 8 | 537 |