Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRCAM SIRNA Assays ›
Gene Symbol:
NRCAM
Gene Name:
neuronal cell adhesion molecule
Gene Aliases:
-
Chromosome Location:
Chr.7: 108147626 - 108456442 on Build GRCh38
UniGene ID:
Hs.21422
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s9715
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRCAM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037132.2
NP_001032209.1
30
3942
NM_001193582.1
NP_001180511.1
31
4043
NM_001193583.1
NP_001180512.1
30
3986
NM_001193584.1
NP_001180513.1
29
3950
NM_005010.4
NP_005001.3
28
3959
XM_005250373.2
XP_005250430.1
33
4322
XM_005250383.2
XP_005250440.1
31
4052
XM_005250385.2
XP_005250442.1
30
4007
XM_006716003.1
XP_006716066.1
34
4334
XM_006716007.1
XP_006716070.1
33
4298
XM_006716012.1
XP_006716075.1
32
4055
XM_006716014.1
XP_006716077.1
31
4019
XM_011516253.1
XP_011514555.1
34
4343
XM_011516255.1
XP_011514557.1
33
4098
XM_011516256.1
XP_011514558.1
33
4331
XM_011516257.1
XP_011514559.1
33
4325
XM_011516258.1
XP_011514560.1
32
4080
XM_011516259.1
XP_011514561.1
33
4313
XM_011516261.1
XP_011514563.1
32
4295
XM_011516262.1
XP_011514564.1
33
4286
XM_011516267.1
XP_011514569.1
32
4064
XM_011516268.2
XP_011514570.1
32
4158
XM_011516269.1
XP_011514571.1
31
4028
XM_011516270.1
XP_011514572.1
30
4016
XM_011516271.1
XP_011514573.1
28
3968
XM_017012236.1
XP_016867725.1
33
4304
XM_017012237.1
XP_016867726.1
32
4286
XM_017012238.1
XP_016867727.1
32
4274
XM_017012239.1
XP_016867728.1
32
4268
XM_017012240.1
XP_016867729.1
32
4265
XM_017012241.1
XP_016867730.1
32
4001
XM_017012242.1
XP_016867731.1
30
3953
XM_017012243.1
XP_016867732.1
33
4181
XM_017012244.1
XP_016867733.1
32
4178
XM_017012245.1
XP_016867734.1
32
4169
XM_017012246.1
XP_016867735.1
31
4025
XM_017012247.1
XP_016867736.1
31
3998
XM_017012248.1
XP_016867737.1
30
3998
XM_017012249.1
XP_016867738.1
30
3995
XM_017012250.1
XP_016867739.1
30
3731
XM_017012251.1
XP_016867740.1
30
3989
XM_017012252.1
XP_016867741.1
29
3986
XM_017012253.1
XP_016867742.1
28
3688
XM_017012254.1
XP_016867743.1
29
3977
XM_017012255.1
XP_016867744.1
30
4032
XM_017012256.1
XP_016867745.1
27
3676
XM_017012257.1
XP_016867746.1
29
3741
XM_017012258.1
XP_016867747.1
29
3760
XM_017012259.1
XP_016867748.1
27
3705
GenBank mRNA
AB002341.1
-
29
3878
AK092330.1
-
3
873
AK127035.1
-
1
2113
AK294195.1
-
4
492
AY528240.1
-
3
469
BC114570.1
-
29
3874
40083055
BC115736.1
-
30
3910
40083062
BX538010.1
-
28
3904
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_013150, U81037, AY574274, AY574277, AY574273, and others.This siRNA also targets the following transcripts in Mouse: NM_176930, NM_001146031, AK122252, AY528242, AJ543321, and others.
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
neuron migration
axonogenesis
axonal fasciculation
synapse assembly
central nervous system development
protein localization
single organismal cell-cell adhesion
neuronal action potential propagation
regulation of axon extension
retinal ganglion cell axon guidance
heterotypic cell-cell adhesion
clustering of voltage-gated sodium channels
positive regulation of neuron differentiation
protein binding
ankyrin binding
protein binding involved in heterotypic cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline