Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133787.2 | NP_598548.1 | 11 | 1033 | ||
GenBank mRNA | AK034772.1 | - | 233 | |||
AK088667.1 | - | 11 | 1046 | |||
AK145248.1 | - | 11 | 1055 | |||
AK166283.1 | - | 11 | 1001 | |||
BC003842.1 | - | 11 | 1033 | 3495021 |