Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101662.1 | NP_001095132.1 | 22 | 2807 | ||
NM_001242361.1 | NP_001229290.1 | 24 | 2406 | |||
NM_002525.2 | NP_002516.2 | 24 | 3011 | |||
XM_005270903.1 | XP_005270960.1 | 22 | 2154 | |||
XM_011541522.1 | XP_011539824.1 | 25 | 3000 | |||
XM_011541523.1 | XP_011539825.1 | 22 | 2371 | |||
XM_011541525.2 | XP_011539827.1 | 24 | 2469 | |||
XM_017001375.1 | XP_016856864.1 | 22 | 2231 | |||
GenBank mRNA | AK055516.1 | - | 22 | 2151 | ||
AK299271.1 | - | 22 | 2254 | |||
AK302616.1 | - | 22 | 2231 | |||
AK312318.1 | - | 22 | 2619 | |||
AY049784.1 | - | 22 | 2522 | |||
AY360265.1 | - | 24 | 2382 | |||
BC008775.2 | - | 22 | 2581 | 3138712 | ||
U64898.1 | - | 22 | 2579 | |||
X93207.1 | - | 24 | 2824 | |||
X93209.1 | - | 22 | 2620 |