Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168270.1 | NP_001161742.1 | 5 | 465 | ||
NM_133794.2 | NP_598555.2 | 9 | 811 | |||
GenBank mRNA | AK004814.1 | - | 9 | 745 | ||
AK030554.1 | - | 9 | 758 | |||
AK088049.1 | - | 9 | 758 | |||
AK145632.1 | - | 9 | 762 | |||
AK155759.1 | - | 9 | 760 | |||
AK165454.1 | - | 9 | 758 | |||
AK167782.1 | - | 9 | 758 | |||
AK170826.1 | - | 9 | 760 | |||
AK171293.1 | - | 9 | 760 | |||
BC023023.1 | - | 5 | 465 | 5362353 | ||
BC079854.1 | - | 9 | 745 | 5717082 |