Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145380.2 | NP_663355.1 | 5 | 561 | ||
GenBank mRNA | AK077480.1 | - | 5 | 562 | ||
AK084738.1 | - | 5 | 580 | |||
AK146222.1 | - | 5 | 593 | |||
AK168066.1 | - | 5 | 578 | |||
AK177627.1 | - | 2 | 130 | |||
BC005598.1 | - | 5 | 561 | 3969335 | ||
BC103795.1 | - | 5 | 561 | 30135815 | ||
BC116787.1 | - | 5 | 558 | 40126106 | ||
BC116789.1 | - | 5 | 558 | 40126108 |