Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145380.2 | NP_663355.1 | 8, 9 | 848 | ||
GenBank mRNA | AK077480.1 | - | 8, 9 | 849 | ||
AK084738.1 | - | 8, 9 | 867 | |||
AK146222.1 | - | 8, 9 | 880 | |||
AK168066.1 | - | 8, 9 | 865 | |||
BC005598.1 | - | 8, 9 | 848 | 3969335 | ||
BC050191.1 | - | 2, 3 | 1580 | |||
BC103795.1 | - | 8, 9 | 848 | 30135815 | ||
BC116787.1 | - | 8, 9 | 845 | 40126106 | ||
BC116789.1 | - | 8, 9 | 845 | 40126108 |