Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133807.1 | NP_598568.1 | 2 | 305 | ||
GenBank mRNA | AK053450.1 | - | 2 | 332 | ||
AK159374.1 | - | 2 | 361 | |||
AK159552.1 | - | 2 | 355 | |||
AK166576.1 | - | 2 | 354 | |||
AK167604.1 | - | 2 | 296 | |||
AK169250.1 | - | 2 | 280 | |||
AK169390.1 | - | 2 | 304 | |||
AK183165.1 | - | 2 | 91 | |||
BC006877.1 | - | 2 | 305 | 2647513 | ||
BC066172.1 | - | 2 | 299 | 30429192 |