Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133810.3 | NP_598571.2 | 6 | 815 | ||
GenBank mRNA | AK014023.1 | - | 1 | 81 | ||
AK029551.1 | - | 6 | 758 | |||
AK035693.1 | - | 6 | 783 | |||
AK041139.1 | - | 2 | 150 | |||
AK076932.1 | - | 6 | 782 | |||
AK083332.1 | - | 6 | 784 | |||
AK153108.1 | - | 6 | 759 | |||
AK165291.1 | - | 6 | 755 | |||
AY092028.1 | - | 6 | 768 | |||
BC006579.1 | - | 6 | 770 | 3586767 |