Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133810.3 | NP_598571.2 | 5 | 726 | ||
GenBank mRNA | AK035693.1 | - | 5 | 694 | ||
AK041139.1 | - | 1 | 61 | |||
AK076932.1 | - | 5 | 693 | |||
AK083332.1 | - | 5 | 695 | |||
AK153108.1 | - | 5 | 670 | |||
AK165291.1 | - | 5 | 666 | |||
AY092028.1 | - | 5 | 679 | |||
BC006579.1 | - | 5 | 681 | 3586767 |