Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D2hgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178882.3 | NP_849213.2 | 7 | 1102 | ||
GenBank mRNA | AK088200.1 | - | 1049 | |||
AK088554.1 | - | 1049 | ||||
AK141691.1 | - | 7 | 1332 | |||
AK170226.1 | - | 1103 | ||||
BC023277.1 | - | 1070 | 4006813 | |||
BC117794.2 | - | 7 | 1040 | 40092173 |