Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slamf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029612.4 | NP_083888.3 | 3 | 556 | ||
GenBank mRNA | AB196824.1 | - | 3 | 419 | ||
AB196825.1 | - | 3 | 419 | |||
AB196826.1 | - | 3 | 419 | |||
AB196827.1 | - | 3 | 419 | |||
AK009505.1 | - | 3 | 512 | |||
AK090041.1 | - | 3 | 547 | |||
BC019477.1 | - | 3 | 495 | 4458767 |