Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcrla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160215.1 | NP_001153687.1 | 3 | 470 | ||
NM_145141.2 | NP_660123.1 | 3 | 467 | |||
GenBank mRNA | AF329487.1 | - | 3 | 434 | ||
AF426462.1 | - | 3 | 427 | |||
AF531424.1 | - | 3 | 456 | |||
AK041572.1 | - | 3 | 447 | |||
AK172370.1 | - | 3 | 460 | |||
BB637253.1 | - | 3 | 463 | |||
BC064708.1 | - | 3 | 470 | 30301438 |