Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166427.1 | NP_001159899.1 | 3 | 986 | ||
NM_001166428.1 | NP_001159900.1 | 4 | 925 | |||
NM_001166429.1 | NP_001159901.1 | 4 | 886 | |||
NM_001166430.1 | NP_001159902.1 | 4 | 1237 | |||
NM_001166431.1 | NP_001159903.1 | 3 | 1176 | |||
NM_001166432.1 | NP_001159904.1 | 4 | 977 | |||
NM_133834.2 | NP_598595.1 | 4 | 868 | |||
GenBank mRNA | AK076478.1 | - | 3 | 826 | ||
AK178627.1 | - | 1 | 132 | |||
BC018185.1 | - | 3 | 811 | 3995067 | ||
BC023793.1 | - | 4 | 863 | |||
BC025481.1 | - | 4 | 845 | 5354000 | ||
BC027003.1 | - | 1 | 445 | 5365491 | ||
BC029163.1 | - | 1 | 629 | 4949890 | ||
BC029764.1 | - | 1 | 452 | 4947521 | ||
BC032197.1 | - | 4 | 864 | |||
BC033483.1 | - | 1 | 627 | 4950131 | ||
BC042689.1 | - | 3 | 1109 | |||
BC047346.1 | - | 3 | 1109 | |||
BC089313.1 | - | 4 | 710 | 6808998 |