Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133838.4 | NP_598599.2 | 4 | 869 | ||
GenBank mRNA | AB239915.1 | - | 4 | 780 | ||
AB239916.1 | - | 4 | 780 | |||
AF173639.1 | - | 4 | 785 | |||
AK147186.1 | - | 4 | 821 | |||
AK149760.1 | - | 4 | 813 | |||
AK159235.1 | - | 4 | 829 | |||
AK159597.1 | - | 4 | 829 | |||
AK163835.1 | - | 4 | 813 | |||
AK166101.1 | - | 4 | 880 | |||
AK169343.1 | - | 4 | 880 | |||
AK178676.1 | - | 3 | 400 | |||
BC007480.1 | - | 4 | 909 | 3493227 | ||
BC012272.1 | - | 4 | 802 | 3485836 |