Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam102a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153560.4 | NP_705788.1 | 8, 9 | 1668 | ||
GenBank mRNA | AK049032.1 | - | 8, 9 | 1672 | ||
AK131145.1 | - | 7, 8 | 3386 | |||
AK157331.1 | - | 8, 9 | 1298 | |||
AK157490.1 | - | 9, 10 | 1090 | |||
AK158435.1 | - | 8, 9 | 1297 | |||
AK165751.1 | - | 8, 9 | 1303 | |||
AK170536.1 | - | 8, 9 | 1696 | |||
AK170847.1 | - | 8, 9 | 1289 | |||
AK170951.1 | - | 8, 9 | 1295 | |||
AK172536.1 | - | 8, 9 | 1295 | |||
AK186631.1 | - | 3, 4 | 344 | |||
BC023752.1 | - | 8, 9 | 1113 | 5354455 | ||
BC031157.1 | - | 8, 9 | 931 | 4976773 |