Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153126.2 | NP_694766.1 | 9, 10 | 1132 | ||
GenBank mRNA | AK036294.1 | - | 9, 10 | 1080 | ||
AK040070.1 | - | 9, 10 | 1087 | |||
AK048552.1 | - | 8, 9 | 957 | |||
AK054029.1 | - | 9, 10 | 1153 | |||
AK087445.1 | - | 9, 10 | 1090 | |||
AK149825.1 | - | 9, 10 | 959 | |||
BC034516.1 | - | 9, 10 | 1132 | 4458024 |