Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153126.2 | NP_694766.1 | 14, 15 | 1743 | ||
GenBank mRNA | AK036294.1 | - | 14, 15 | 1639 | ||
AK040070.1 | - | 14, 15 | 1698 | |||
AK048552.1 | - | 13, 14 | 1568 | |||
AK054029.1 | - | 14, 15 | 1764 | |||
AK149825.1 | - | 14, 15 | 1570 | |||
AK184974.1 | - | 2, 3 | 266 | |||
BC034516.1 | - | 14, 15 | 1743 | 4458024 | ||
BC047436.1 | - | 2, 3 | 276 |