Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpcat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207206.2 | NP_997089.1 | 3 | 500 | ||
GenBank mRNA | BC046276.1 | - | 2 | 250 | ||
BC068131.1 | - | 3 | 500 | 6414152 | ||
BC080829.1 | - | 3 | 467 | 30435722 |