Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290733.1 | NP_001277662.1 | 27 | 3550 | ||
NM_001290734.1 | NP_001277663.1 | 25 | 3217 | |||
NM_145525.3 | NP_663500.2 | 27 | 3334 | |||
GenBank mRNA | AK037703.1 | - | 25 | 3168 | ||
AK044411.1 | - | 27 | 3521 | |||
AK048018.1 | - | 2 | 144 | |||
AK220498.1 | - | 27 | 3281 | |||
BC022908.1 | - | 21 | 2539 | 5043261 |