Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207207.1 | NP_997090.1 | 2 | 254 | ||
GenBank mRNA | AK131626.1 | - | 2 | 264 | ||
AK169837.1 | - | 2 | 267 | |||
BC048517.1 | - | 2 | 254 | 6706680 |