Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139308.2 | NP_647469.2 | 2 | 846 | ||
GenBank mRNA | AF244543.1 | - | 1 | 203 | ||
AK044519.1 | - | 2 | 342 | |||
AK083125.1 | - | 2 | 373 | |||
AK143699.1 | - | 2 | 847 | |||
AK199440.1 | - | 1 | 22 | |||
AK208068.1 | - | 1 | 22 | |||
AK217776.1 | - | 22 | ||||
AK218007.1 | - | 1 | 22 | |||
AK219177.1 | - | 1 | 22 | |||
BC017524.1 | - | 2 | 836 | 4527790 | ||
BC080747.1 | - | 2 | 817 | 4237000 |