Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssx2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253768.1 | NP_001240697.1 | 6, 7 | 1082 | ||
NM_001253769.1 | NP_001240698.1 | 5, 6 | 978 | |||
NM_001253770.1 | NP_001240699.1 | 5, 6 | 827 | |||
NM_138744.3 | NP_620083.1 | 5, 6 | 827 | |||
GenBank mRNA | AF532969.1 | - | 4, 5 | 622 | ||
AK031356.1 | - | 6, 7 | 806 | |||
AK043865.1 | - | 5, 6 | 931 | |||
AK049080.1 | - | 5, 6 | 979 | |||
AK077112.1 | - | 6, 7 | 867 | |||
AK173064.1 | - | 5, 6 | 814 | |||
BC021749.1 | - | 5, 6 | 782 | 4165430 | ||
BC031527.1 | - | 5, 6 | 790 | 4008624 |