Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133847.3 | NP_598608.2 | 8 | 1063 | ||
GenBank mRNA | AB093220.2 | - | 7 | 807 | ||
AK035622.1 | - | 8 | 1025 | |||
AK047497.1 | - | 8 | 790 | |||
AK048158.1 | - | 8 | 821 | |||
AK083541.1 | - | 8 | 1063 | |||
AK087048.1 | - | 8 | 816 | |||
AK088012.1 | - | 8 | 819 | |||
AK162435.1 | - | 8 | 824 | |||
AK171487.1 | - | 8 | 821 | |||
AK194933.1 | - | 3 | 189 | |||
BC110309.1 | - | 8 | 819 | 5324218 | ||
BC116320.2 | - | 8 | 1015 | 40096002 |