Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133847.3 | NP_598608.2 | 16 | 1922 | ||
GenBank mRNA | AB093220.2 | - | 15 | 1666 | ||
AK035622.1 | - | 16 | 1884 | |||
AK048158.1 | - | 16 | 1680 | |||
AK083541.1 | - | 16 | 1922 | |||
AK087048.1 | - | 16 | 1675 | |||
AK162435.1 | - | 16 | 1683 | |||
AK171487.1 | - | 16 | 1680 | |||
BC006741.1 | - | 2 | 359 | |||
BC071208.1 | - | 3 | 741 | |||
BC110309.1 | - | 16 | 1678 | 5324218 | ||
BC116320.2 | - | 16 | 1874 | 40096002 |