Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133847.3 | NP_598608.2 | 14 | 1747 | ||
GenBank mRNA | AB093220.2 | - | 13 | 1491 | ||
AK035622.1 | - | 14 | 1709 | |||
AK048158.1 | - | 14 | 1505 | |||
AK083541.1 | - | 14 | 1747 | |||
AK087048.1 | - | 14 | 1500 | |||
AK088012.1 | - | 14 | 1503 | |||
AK162435.1 | - | 14 | 1508 | |||
AK171487.1 | - | 14 | 1505 | |||
BC071208.1 | - | 1 | 566 | |||
BC110309.1 | - | 14 | 1503 | 5324218 | ||
BC116320.2 | - | 14 | 1699 | 40096002 |