Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146207.2 | NP_666319.2 | 12 | 1276 | ||
GenBank mRNA | AK144802.1 | - | 12 | 1259 | ||
AK150237.1 | - | 12 | 1255 | |||
AK168215.1 | - | 12 | 1258 | |||
AK170722.1 | - | 12 | 1258 | |||
BC010211.1 | - | 12 | 1254 | 3969515 | ||
BC024113.1 | - | 8 | 748 | 5097944 | ||
BC027006.1 | - | 8 | 796 |