Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146207.2 | NP_666319.2 | 19 | 2139 | ||
GenBank mRNA | AK003679.1 | - | 3 | 340 | ||
AK005426.1 | - | 4 | 442 | |||
AK144802.1 | - | 19 | 2122 | |||
AK168215.1 | - | 19 | 2121 | |||
AK170722.1 | - | 19 | 2121 | |||
AK183542.1 | - | 3 | 304 | |||
AK190734.1 | - | 2 | 193 | |||
AK194132.1 | - | 2 | 222 | |||
BC010211.1 | - | 19 | 2117 | 3969515 | ||
BC024113.1 | - | 15 | 1611 | 5097944 | ||
BC024667.1 | - | 1 | 125 | |||
BC027006.1 | - | 15 | 1659 |