Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080968.1 | NP_001074437.1 | 7, 8 | 601 | ||
NM_133852.2 | NP_598613.2 | 8, 9 | 682 | |||
GenBank mRNA | AK076370.1 | - | 3, 4 | 222 | ||
AK077516.1 | - | 7, 8 | 602 | |||
AK220509.1 | - | 8, 9 | 610 | |||
BC011407.1 | - | 6, 7 | 402 | 3596079 |