Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080968.1 | NP_001074437.1 | 11 | 889 | ||
NM_133852.2 | NP_598613.2 | 12 | 970 | |||
GenBank mRNA | AK076370.1 | - | 6 | 588 | ||
AK077516.1 | - | 11 | 891 | |||
AK220509.1 | - | 12 | 898 | |||
BC011407.1 | - | 10 | 690 | 3596079 |