Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133862.1 | NP_598623.1 | 8 | 1127 | ||
GenBank mRNA | AK013887.1 | - | 8 | 1170 | ||
AK149375.1 | - | 8 | 1163 | |||
AK149448.1 | - | 8 | 1164 | |||
BC019506.1 | - | 8 | 1127 | 4193631 | ||
BC019828.1 | - | 8 | 1137 | 5123696 |