Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133862.1 | NP_598623.1 | 8 | 1043 | ||
GenBank mRNA | AF413206.1 | - | 4 | 524 | ||
AK013887.1 | - | 8 | 1086 | |||
AK149375.1 | - | 8 | 1079 | |||
AK149448.1 | - | 8 | 1080 | |||
BC019506.1 | - | 8 | 1043 | 4193631 | ||
BC019828.1 | - | 8 | 1053 | 5123696 |