Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933434E20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287087.1 | NP_001274016.1 | 6, 7 | 746 | ||
NM_025762.3 | NP_080038.1 | 4, 5 | 601 | |||
NM_027500.4 | NP_081776.2 | 4, 5 | 601 | |||
GenBank mRNA | AK017833.1 | - | 4, 5 | 569 | ||
AK034749.1 | - | 4, 5 | 564 | |||
AK051238.1 | - | 4, 5 | 494 | |||
AK077215.1 | - | 6, 7 | 747 | |||
AK150324.1 | - | 4, 5 | 552 | |||
AK150423.1 | - | 4, 5 | 552 | |||
AK150831.1 | - | 4, 5 | 552 | |||
AK151002.1 | - | 4, 5 | 551 | |||
AK151263.1 | - | 4, 5 | 550 | |||
AK214769.1 | - | 4, 5 | 352 | |||
AY533135.1 | - | 4, 5 | 349 | |||
BC027190.1 | - | 4, 5 | 461 | 3257131 | ||
BG091444.1 | - | 2, 3 | 196 |