Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps8l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133867.2 | NP_598628.1 | 16 | 1669 | ||
GenBank mRNA | AK136584.1 | - | 16 | 1666 | ||
AY074932.1 | - | 16 | 1685 | |||
BC014734.1 | - | 16 | 1657 | 4218079 |